siRNA Details
Gene Symbol: SHANK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012309.4 | NP_036441.2 | 4 | 540 | ||
XM_017017387.1 | XP_016872876.1 | 5 | 645 | |||
XM_017017388.1 | XP_016872877.1 | 5 | 667 | |||
XM_017017389.1 | XP_016872878.1 | 5 | 840 | |||
GenBank mRNA | AB208025.1 | - | 4 | 506 | ||
AK096045.1 | - | 7 | 717 | |||
DQ152234.1 | - | 3 | 428 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
