siRNA Details
Gene Symbol: ADCY10P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK126092.1 | - | 4 | 864 | ||
AK128294.1 | - | 2 | 194 | |||
BC126399.1 | - | 3 | 244 | |||
BX537595.1 | - | 15 | 4356 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
