siRNA Details
Gene Symbol: ANKFN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006721728.3 | XP_006721791.1 | 17, 18 | 2705 | ||
XM_011524428.2 | XP_011522730.1 | 18, 19 | 2482 | |||
XM_011524429.2 | XP_011522731.1 | 18, 19 | 2639 | |||
XM_011524430.2 | XP_011522732.1 | 19, 20 | 2605 | |||
XM_011524431.2 | XP_011522733.1 | 18, 19 | 2537 | |||
XM_011524433.2 | XP_011522735.1 | 12, 13 | 1895 | |||
XM_011524434.2 | XP_011522736.1 | 9, 10 | 1342 | |||
XM_017024263.1 | XP_016879752.1 | 20, 21 | 2699 | |||
XM_017024264.1 | XP_016879753.1 | 20, 21 | 3684 | |||
XM_017024265.1 | XP_016879754.1 | 20, 21 | 2893 | |||
XM_017024266.1 | XP_016879755.1 | 18, 19 | 2654 | |||
XM_017024267.1 | XP_016879756.1 | 17, 18 | 2506 | |||
XM_017024268.1 | XP_016879757.1 | 19, 20 | 3590 | |||
XM_017024269.1 | XP_016879758.1 | 17, 18 | 2779 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
