siRNA Details
Gene Symbol: ANKFN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006721728.3 | XP_006721791.1 | 19 | 3033 | ||
XM_011524428.2 | XP_011522730.1 | 20 | 2810 | |||
XM_011524429.2 | XP_011522731.1 | 20 | 2967 | |||
XM_011524430.2 | XP_011522732.1 | 21 | 2933 | |||
XM_011524431.2 | XP_011522733.1 | 20 | 2865 | |||
XM_011524433.2 | XP_011522735.1 | 14 | 2223 | |||
XM_011524434.2 | XP_011522736.1 | 11 | 1670 | |||
XM_017024263.1 | XP_016879752.1 | 22 | 3027 | |||
XM_017024264.1 | XP_016879753.1 | 22 | 4012 | |||
XM_017024265.1 | XP_016879754.1 | 22 | 3221 | |||
XM_017024266.1 | XP_016879755.1 | 20 | 2982 | |||
XM_017024267.1 | XP_016879756.1 | 19 | 2834 | |||
XM_017024268.1 | XP_016879757.1 | 21 | 3918 | |||
XM_017024269.1 | XP_016879758.1 | 19 | 3107 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
