siRNA Details
Gene Symbol: PMS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000535.6 | NP_000526.2 | 15 | 2840 | ||
NM_001322003.1 | NP_001308932.1 | 15 | 2835 | |||
NM_001322004.1 | NP_001308933.1 | 14 | 2700 | |||
NM_001322005.1 | NP_001308934.1 | 15 | 2952 | |||
NM_001322006.1 | NP_001308935.1 | 14 | 2684 | |||
NM_001322007.1 | NP_001308936.1 | 14 | 2737 | |||
NM_001322008.1 | NP_001308937.1 | 13 | 2597 | |||
NM_001322009.1 | NP_001308938.1 | 15 | 2980 | |||
NM_001322010.1 | NP_001308939.1 | 13 | 2544 | |||
NM_001322011.1 | NP_001308940.1 | 15 | 2791 | |||
NM_001322012.1 | NP_001308941.1 | 15 | 2786 | |||
NM_001322013.1 | NP_001308942.1 | 14 | 2667 | |||
NM_001322014.1 | NP_001308943.1 | 15 | 2873 | |||
NM_001322015.1 | NP_001308944.1 | 15 | 3010 | |||
NR_136154.1 | 15 | 2797 | ||||
XM_006715744.3 | XP_006715807.1 | 8 | 1916 | |||
XM_017012342.1 | XP_016867831.1 | 14 | 2665 | |||
GenBank mRNA | AK125607.1 | - | 1 | 41 | ||
BQ050630.1 | - | 3 | 534 | |||
DB548617.1 | - | 1 | 64 |
Gene Symbol: PMS2CL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002217.1 | 6 | 1694 | |||
GenBank mRNA | AB116525.1 | - | 6 | 1694 | ||
BC041364.1 | - | 13 | 2449 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
