siRNA Details
Gene Symbol: PLET1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145024.1 | NP_001138496.1 | 2 | 569 | ||
GenBank mRNA | AY364431.1 | - | 2 | 298 | ||
AY364434.1 | - | 1 | 115 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
