siRNA Details
Gene Symbol: RBFOX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001082575.2 | NP_001076044.1 | 7 | 900 | ||
XM_011524359.1 | XP_011522661.1 | 7 | 982 | |||
XM_011524360.1 | XP_011522662.1 | 8 | 1171 | |||
XM_011524362.2 | XP_011522664.1 | 7 | 979 | |||
XM_011524363.2 | XP_011522665.1 | 7 | 982 | |||
XM_011524365.2 | XP_011522667.1 | 7 | 982 | |||
XM_011524366.2 | XP_011522668.1 | 7 | 979 | |||
XM_011524367.2 | XP_011522669.1 | 7 | 982 | |||
XM_017024208.1 | XP_016879697.1 | 6 | 942 | |||
XM_017024209.1 | XP_016879698.1 | 7 | 982 | |||
XM_017024210.1 | XP_016879699.1 | 7 | 982 | |||
XM_017024211.1 | XP_016879700.1 | 7 | 982 | |||
GenBank mRNA | AK293617.1 | - | 7 | 900 | ||
AK293905.1 | - | 7 | 763 | |||
BX452143.2 | - | 3 | 422 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
