siRNA Details
Gene Symbol: RBFOX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001082575.2 | NP_001076044.1 | 14 | 1438 | ||
XM_011524359.1 | XP_011522661.1 | 14 | 1661 | |||
XM_011524360.1 | XP_011522662.1 | 15 | 1850 | |||
XM_011524362.2 | XP_011522664.1 | 14 | 1658 | |||
XM_011524363.2 | XP_011522665.1 | 14 | 1661 | |||
XM_011524365.2 | XP_011522667.1 | 14 | 1520 | |||
XM_011524366.2 | XP_011522668.1 | 14 | 1517 | |||
XM_011524367.2 | XP_011522669.1 | 13 | 1427 | |||
XM_017024208.1 | XP_016879697.1 | 13 | 1621 | |||
XM_017024209.1 | XP_016879698.1 | 13 | 1568 | |||
XM_017024210.1 | XP_016879699.1 | 14 | 1520 | |||
XM_017024211.1 | XP_016879700.1 | 13 | 1427 | |||
GenBank mRNA | AK124644.1 | - | 5 | 1120 | ||
AK128131.1 | - | 5 | 3021 | |||
AK293617.1 | - | 14 | 1438 | |||
AK293905.1 | - | 14 | 1301 | |||
BM714144.1 | - | 7 | 519 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
