siRNA Details
Gene Symbol: CATSPER4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198137.1 | NP_937770.1 | 3 | 369 | ||
XM_011541432.2 | XP_011539734.1 | 3 | 476 | |||
XM_011541433.2 | XP_011539735.1 | 3 | 1035 | |||
GenBank mRNA | BC128138.1 | - | 3 | 368 | 40112485 | |
BC128139.1 | - | 3 | 369 | 40112493 | ||
BC144663.1 | - | 3 | 436 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
