siRNA Details
Gene Symbol: NFU1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002755.2 | NP_001002755.1 | 3 | 465 | ||
NM_015700.3 | NP_056515.2 | 3 | 555 | |||
XM_017003808.1 | XP_016859297.1 | 3 | 268 | |||
GenBank mRNA | AF132967.1 | - | 3 | 264 | ||
AJ132584.1 | - | 3 | 343 | |||
AK300700.1 | - | 3 | 274 | |||
AK314004.1 | - | 3 | 298 | |||
AY286306.1 | - | 3 | 312 | |||
AY286307.1 | - | 3 | 402 | |||
AY335194.1 | - | 3 | 458 | |||
BC113692.1 | - | 3 | 427 | 8322744 | ||
BC113694.1 | - | 3 | 427 | 8322746 | ||
BM846341.1 | - | 3 | 465 | |||
BU507991.1 | - | 3 | 270 | |||
BX538347.1 | - | 3 | 330 | |||
DB496651.2 | - | 3 | 451 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
