siRNA Details
Gene Symbol: HELT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300781.1 | NP_001287710.1 | 4 | 522 | ||
NM_001300782.1 | NP_001287711.1 | 4 | 519 | |||
XM_017008186.1 | XP_016863675.1 | 3 | 565 | |||
GenBank mRNA | BC140795.1 | - | 4 | 519 | 9021663 | |
BC144567.1 | - | 4 | 522 | 9053095 | ||
BC144568.1 | - | 3 | 774 | |||
BC144569.1 | - | 4 | 519 | 9053097 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
