siRNA Details
Gene Symbol: NCR3LG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202439.2 | NP_001189368.1 | 3 | 938 | ||
XM_011520072.2 | XP_011518374.1 | 3 | 2892 | |||
XM_011520073.2 | XP_011518375.1 | 3 | 2638 | |||
XM_011520074.2 | XP_011518376.1 | 3 | 1255 | |||
XM_011520075.2 | XP_011518377.1 | 3 | 2544 | |||
GenBank mRNA | BC136797.1 | - | 3 | 801 | ||
BC136800.1 | - | 3 | 801 | |||
BX537685.1 | - | 3 | 930 | |||
CR749521.1 | - | 3 | 938 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
