siRNA Details
Gene Symbol: NCR3LG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202439.2 | NP_001189368.1 | 1, 2 | 278 | ||
GenBank mRNA | BC136797.1 | - | 1, 2 | 141 | ||
BC136800.1 | - | 1, 2 | 141 | |||
BX537685.1 | - | 1, 2 | 270 | |||
CR749521.1 | - | 1, 2 | 278 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
