siRNA Details
Gene Symbol: EPHA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080448.2 | NP_001073917.2 | 3 | 536 | ||
NM_001278301.1 | NP_001265230.1 | 3 | 536 | |||
XM_006713592.3 | XP_006713655.1 | 3 | 536 | |||
XM_011512705.2 | XP_011511007.1 | 3 | 536 | |||
XM_011512706.2 | XP_011511008.1 | 3 | 536 | |||
XM_011512707.2 | XP_011511009.1 | 3 | 536 | |||
XM_017006210.1 | XP_016861699.1 | 3 | 536 | |||
XM_017006211.1 | XP_016861700.1 | 4 | 561 | |||
XM_017006212.1 | XP_016861701.1 | 3 | 230 | |||
XM_017006213.1 | XP_016861702.1 | 3 | 588 | |||
XM_017006219.1 | XP_016861708.1 | 3 | 536 | |||
GenBank mRNA | AK092565.1 | - | 3 | 531 | ||
AY358738.1 | - | 3 | 337 | |||
DA385338.1 | - | 3 | 536 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
