siRNA Details
Gene Symbol: EPHA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080448.2 | NP_001073917.2 | 5 | 1619 | ||
XM_006713592.3 | XP_006713655.1 | 5 | 1619 | |||
XM_017006210.1 | XP_016861699.1 | 5 | 1619 | |||
XM_017006211.1 | XP_016861700.1 | 6 | 1644 | |||
XM_017006212.1 | XP_016861701.1 | 5 | 1313 | |||
XM_017006213.1 | XP_016861702.1 | 5 | 1671 | |||
XM_017006214.1 | XP_016861703.1 | 4 | 939 | |||
XM_017006219.1 | XP_016861708.1 | 5 | 1619 | |||
GenBank mRNA | AK092565.1 | - | 5 | 1614 | ||
DN991634.1 | - | 1 | 82 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
