siRNA Details
Gene Symbol: SMIM15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048249.3 | NP_001041714.1 | 3 | 803 | ||
GenBank mRNA | BC014242.2 | - | 3 | 657 | ||
BC047489.2 | - | 3 | 684 | |||
BC066550.1 | - | 3 | 782 | |||
BC103711.1 | - | 3 | 758 | |||
BX538015.1 | - | 3 | 798 | |||
BX648459.1 | - | 3 | 775 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
