siRNA Details
Gene Symbol: SMIM15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048249.3 | NP_001041714.1 | 3 | 2766 | ||
GenBank mRNA | BX538015.1 | - | 3 | 2765 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
