siRNA Details
Gene Symbol: AFF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014423.3 | NP_055238.1 | 21 | 8870 | ||
XM_005271963.4 | XP_005272020.1 | 22 | 8934 | |||
XM_005271964.4 | XP_005272021.1 | 15 | 7582 | |||
XM_006714587.3 | XP_006714650.1 | 20 | 8783 | |||
GenBank mRNA | AF173886.1 | - | 1 | 881 | ||
AK126818.1 | - | 1 | 760 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
