siRNA Details
Gene Symbol: KIAA0825 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145678.1 | NP_001139150.1 | 17 | 3529 | ||
XM_006714588.2 | XP_006714651.1 | 17 | 3544 | |||
XM_011543324.2 | XP_011541626.1 | 17 | 3544 | |||
XM_011543325.2 | XP_011541627.1 | 18 | 3610 | |||
XM_011543327.2 | XP_011541629.1 | 17 | 3460 | |||
XM_011543328.2 | XP_011541630.1 | 18 | 3665 | |||
XM_011543329.2 | XP_011541631.1 | 16 | 3380 | |||
XM_011543330.2 | XP_011541632.1 | 17 | 3544 | |||
XM_011543331.1 | XP_011541633.1 | 17 | 3544 | |||
XM_011543332.1 | XP_011541634.1 | 17 | 3544 | |||
XM_011543333.2 | XP_011541635.1 | 17 | 3544 | |||
XM_011543334.2 | XP_011541636.1 | 17 | 3544 | |||
XM_011543335.2 | XP_011541637.1 | 14 | 3077 | |||
XM_017009373.1 | XP_016864862.1 | 17 | 3544 | |||
XM_017009374.1 | XP_016864863.1 | 17 | 3544 | |||
XM_017009375.1 | XP_016864864.1 | 18 | 3609 | |||
GenBank mRNA | AK130941.1 | - | 1 | 154 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
