siRNA Details
Gene Symbol: STMND1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190766.1 | NP_001177695.1 | 2 | 249 | ||
GenBank mRNA | DT932725.1 | - | 1 | 89 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
