siRNA Details
Gene Symbol: NANOS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098622.2 | NP_001092092.1 | 1 | 33 | ||
XM_011527964.2 | XP_011526266.1 | 1 | 899 | |||
GenBank mRNA | BC101209.2 | - | 1 | 36 | 40021950 | |
BC101210.1 | - | 1 | 36 | |||
DN993610.1 | - | 1 | 154 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
