siRNA Details
Gene Symbol: C9orf153 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276366.1 | NP_001263295.1 | 3 | 322 | ||
NM_001276367.1 | NP_001263296.1 | 4 | 401 | |||
NM_001276368.1 | NP_001263297.1 | 4 | 340 | |||
GenBank mRNA | BC108665.1 | - | 4 | 379 | 5164660 | |
BG772848.1 | - | 3 | 327 | |||
BI827110.1 | - | 4 | 341 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
