siRNA Details
Gene Symbol: KIF18B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001264573.1 | NP_001251503.1 | 7 | 1117 | ||
NM_001265577.1 | NP_001252506.1 | 7 | 1117 | |||
XM_011524385.2 | XP_011522687.1 | 7 | 1418 | |||
XM_011524386.2 | XP_011522688.1 | 7 | 1152 | |||
XM_011524387.2 | XP_011522689.1 | 7 | 1419 | |||
XM_011524388.2 | XP_011522690.1 | 7 | 1418 | |||
XM_011524389.2 | XP_011522691.1 | 7 | 1418 | |||
XM_011524390.2 | XP_011522692.1 | 6 | 1314 | |||
XM_011524391.2 | XP_011522693.1 | 7 | 1421 | |||
GenBank mRNA | BC136590.1 | - | 6 | 1043 | 9020580 | |
BC144271.1 | - | 6 | 1043 | 9052791 | ||
GU808359.1 | - | 6 | 1046 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
