siRNA Details
Gene Symbol: CLEC2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080511.2 | NP_001073980.2 | 5 | 861 | ||
XM_017011770.1 | XP_016867259.1 | 5 | 812 | |||
GenBank mRNA | AK057548.1 | - | 3 | 1764 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
