siRNA Details
Gene Symbol: HLA-DRB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF017440.1 | - | 2 | 168 | ||
AF291675.1 | - | 2 | 181 | |||
AF436097.1 | - | 1 | 58 | |||
AJ297586.2 | - | 2 | 181 | |||
AY961062.1 | - | 2 | 181 | |||
AY961063.1 | - | 2 | 181 | |||
AY961068.1 | - | 2 | 181 | |||
BC024269.1 | - | 2 | 224 | 4852401 | ||
L42143.1 | - | 2 | 181 | |||
M17381.1 | - | 1 | 94 | |||
X02902.1 | - | 2 | 237 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
