siRNA Details
Gene Symbol: KIR3DS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083539.2 | NP_001077008.1 | 9 | 1185 | ||
NM_001282170.1 | NP_001269099.1 | 8 | 900 | |||
NM_001282171.1 | NP_001269100.1 | 7 | 864 | |||
GenBank mRNA | AF022044.1 | - | 9 | 1161 | ||
AJ417558.3 | - | 1097 | ||||
AK301140.1 | - | 9 | 1185 | |||
BC098107.1 | - | 7 | 838 | 40026977 | ||
BC098288.1 | - | 9 | 1159 | 40026980 | ||
BC105679.1 | - | 9 | 1159 | 40004313 | ||
EF370406.1 | - | 8 | 1108 | |||
L76661.1 | - | 9 | 1158 | |||
U73396.1 | - | 9 | 1148 | |||
X97232.1 | - | 8 | 863 | |||
X97233.1 | - | 9 | 1148 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
