siRNA Details
Gene Symbol: KIR3DS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083539.2 | NP_001077008.1 | 9 | 1181 | ||
NM_001282170.1 | NP_001269099.1 | 8 | 896 | |||
NM_001282171.1 | NP_001269100.1 | 7 | 860 | |||
GenBank mRNA | AF022044.1 | - | 9 | 1157 | ||
AJ417558.3 | - | 1093 | ||||
AK301140.1 | - | 9 | 1181 | |||
BC098107.1 | - | 7 | 834 | 40026977 | ||
BC098288.1 | - | 9 | 1155 | 40026980 | ||
BC105679.1 | - | 9 | 1155 | 40004313 | ||
EF370406.1 | - | 8 | 1104 | |||
L76661.1 | - | 9 | 1154 | |||
U73396.1 | - | 9 | 1144 | |||
X97232.1 | - | 8 | 859 | |||
X97233.1 | - | 9 | 1144 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
