siRNA Details
Gene Symbol: CPSF4L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001129885.1 | NP_001123357.1 | 4 | 397 | ||
XM_011525115.2 | XP_011523417.1 | 4 | 2295 | |||
GenBank mRNA | BC157870.1 | - | 4 | 397 | 9057089 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
