siRNA Details
Gene Symbol: TCTEX1D4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013632.3 | NP_001013654.1 | 2 | 1023 | ||
XM_006710614.3 | XP_006710677.1 | 3 | 708 | |||
XM_011541375.2 | XP_011539677.1 | 3 | 758 | |||
XM_011541376.2 | XP_011539678.1 | 3 | 614 | |||
XM_017001187.1 | XP_016856676.1 | 2 | 657 | |||
GenBank mRNA | BC092499.1 | - | 2 | 1023 | 30531412 | |
DQ132441.1 | - | 2 | 1023 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
