siRNA Details
Gene Symbol: LRRIQ4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080460.1 | NP_001073929.1 | 4 | 1350 | ||
XM_005247421.2 | XP_005247478.1 | 5 | 1461 | |||
XM_006713613.3 | XP_006713676.1 | 5 | 1709 | |||
XM_011512748.1 | XP_011511050.1 | 5 | 1460 | |||
XM_011512749.1 | XP_011511051.1 | 4 | 1287 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
