siRNA Details
Gene Symbol: C11orf96 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145033.1 | NP_001138505.1 | 1 | 1050 | ||
GenBank mRNA | BC052560.1 | - | 1 | 1016 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
