siRNA Details
Gene Symbol: VMAC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017921.3 | NP_001017921.1 | 2 | 826 | ||
GenBank mRNA | AK124162.1 | - | 2 | 793 | ||
BC110802.1 | - | 2 | 793 | 5108486 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
