siRNA Details
Gene Symbol: S100A13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024210.1 | NP_001019381.1 | 5 | 704 | ||
NM_001024211.1 | NP_001019382.1 | 3 | 420 | |||
NM_001024212.1 | NP_001019383.1 | 3 | 391 | |||
NM_001024213.1 | NP_001019384.1 | 3 | 359 | |||
NM_005979.2 | NP_005970.1 | 4 | 496 | |||
XM_005245434.3 | XP_005245491.1 | 5 | 611 | |||
XM_011509862.2 | XP_011508164.1 | 7 | 911 | |||
XM_011509863.2 | XP_011508165.1 | 7 | 878 | |||
XM_011509864.1 | XP_011508166.1 | 5 | 701 | |||
XM_017002034.1 | XP_016857523.1 | 6 | 794 | |||
XM_017002035.1 | XP_016857524.1 | 6 | 795 | |||
XM_017002036.1 | XP_016857525.1 | 3 | 528 | |||
GenBank mRNA | AK097132.1 | - | 5 | 2000 | ||
AY987392.1 | - | 2 | 197 | |||
BC000632.2 | - | 2 | 269 | 3343203 | ||
BC068064.1 | - | 2 | 255 | 6074799 | ||
BC070291.1 | - | 4 | 496 | 6713909 | ||
BM762088.1 | - | 3 | 393 | |||
BM913375.1 | - | 563 | ||||
BT006724.1 | - | 2 | 200 | |||
CR542149.1 | - | 2 | 200 | |||
X99920.1 | - | 3 | 273 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
