siRNA Details
Gene Symbol: MSL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012241.1 | NP_001012241.1 | 4 | 821 | ||
XM_005257298.4 | XP_005257355.1 | 3 | 2473 | |||
XM_005257299.4 | XP_005257356.3 | 3 | 2473 | |||
GenBank mRNA | AK297619.1 | - | 1 | 142 | ||
AL049450.1 | - | 2 | 632 | |||
BC118997.1 | - | 4 | 744 | 40084759 | ||
BC122543.1 | - | 4 | 744 | 40084757 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
