siRNA Details
Gene Symbol: BEND3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080450.2 | NP_001073919.1 | 5 | 2663 | ||
XM_005267079.3 | XP_005267136.1 | 4 | 2122 | |||
XM_005267080.3 | XP_005267137.1 | 4 | 2441 | |||
XM_011536005.2 | XP_011534307.1 | 5 | 2152 | |||
XM_017011138.1 | XP_016866627.1 | 5 | 2692 | |||
GenBank mRNA | AB046773.1 | - | 1 | 1540 | ||
BC131628.1 | - | 5 | 2663 | 40118705 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
