siRNA Details
Gene Symbol: SLC35F4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206920.1 | NP_001193849.1 | 7 | 1225 | ||
NM_001306087.1 | NP_001293016.1 | 7 | 1228 | |||
XM_011536720.2 | XP_011535022.1 | 9 | 5007 | |||
XM_011536721.2 | XP_011535023.1 | 9 | 5004 | |||
XM_011536723.2 | XP_011535025.1 | 7 | 1493 | |||
XM_011536724.2 | XP_011535026.1 | 7 | 2098 | |||
XM_011536725.1 | XP_011535027.1 | 8 | 1417 | |||
XM_017021258.1 | XP_016876747.1 | 8 | 1413 | |||
XM_017021260.1 | XP_016876749.1 | 8 | 1420 | |||
GenBank mRNA | BC101318.1 | - | 7 | 1166 | ||
BC101319.1 | - | 9 | 2425 | |||
BC101320.1 | - | 7 | 1172 | |||
BC101321.1 | - | 7 | 1172 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
