siRNA Details
Gene Symbol: CFAP46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001200049.2 | NP_001186978.2 | 28 | 3801 | ||
GenBank mRNA | DV080629.1 | - | 1 | 82 | ||
DY654537.1 | - | 2 | 261 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
