siRNA Details
Gene Symbol: FAM47E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136570.2 | NP_001130042.1 | 3 | 568 | ||
NM_001242936.1 | NP_001229865.1 | 3 | 596 | |||
GenBank mRNA | AK124936.1 | - | 3 | 596 | ||
AK310457.1 | - | 2 | 222 | |||
CR627383.1 | - | 2 | 189 | |||
DB081518.1 | - | 2 | 222 |
Gene Symbol: FAM47E-STBD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242939.1 | NP_001229868.1 | 3 | 568 | ||
GenBank mRNA | AK092277.1 | - | 3 | 547 | ||
AK296418.1 | - | 2 | 189 | |||
AK316039.1 | - | 2 | 190 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
