siRNA Details
Gene Symbol: FAM47E-STBD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242939.1 | NP_001229868.1 | 5, 6 | 920 | ||
GenBank mRNA | AK092277.1 | - | 5, 6 | 899 |
Gene Symbol: FAM47E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136570.2 | NP_001130042.1 | 5, 6 | 920 | ||
NM_001242936.1 | NP_001229865.1 | 5, 6 | 993 | |||
GenBank mRNA | AK124936.1 | - | 5, 6 | 993 | ||
BM713546.1 | - | 1, 2 | 165 | |||
CR627383.1 | - | 4, 5 | 542 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
