siRNA Details
Gene Symbol: ZNF292 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015021.1 | NP_055836.1 | 7 | 996 | ||
XM_005248697.4 | XP_005248754.1 | 7 | 810 | |||
XM_017010575.1 | XP_016866064.1 | 8 | 4187 | |||
XM_017010576.1 | XP_016866065.1 | 9 | 1713 | |||
XM_017010577.1 | XP_016866066.1 | 7 | 4042 | |||
XM_017010578.1 | XP_016866067.1 | 9 | 6695 | |||
XM_017010579.1 | XP_016866068.1 | 9 | 3721 | |||
XM_017010580.1 | XP_016866069.1 | 9 | 1142 | |||
XM_017010581.1 | XP_016866070.1 | 8 | 1128 | |||
XM_017010582.1 | XP_016866071.1 | 7 | 3838 | |||
XM_017010583.1 | XP_016866072.1 | 8 | 1435 | |||
XM_017010584.1 | XP_016866073.1 | 7 | 5811 | |||
GenBank mRNA | BX537746.1 | - | 7 | 971 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
