siRNA Details
Gene Symbol: ZNF292 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015021.1 | NP_055836.1 | 8 | 3362 | ||
XM_005248697.4 | XP_005248754.1 | 8 | 3176 | |||
XM_011535625.2 | XP_011533927.1 | 7 | 3182 | |||
XM_017010575.1 | XP_016866064.1 | 9 | 6553 | |||
XM_017010576.1 | XP_016866065.1 | 10 | 4079 | |||
XM_017010577.1 | XP_016866066.1 | 8 | 6408 | |||
XM_017010578.1 | XP_016866067.1 | 10 | 9061 | |||
XM_017010579.1 | XP_016866068.1 | 10 | 6087 | |||
XM_017010580.1 | XP_016866069.1 | 10 | 3508 | |||
XM_017010581.1 | XP_016866070.1 | 9 | 3494 | |||
XM_017010582.1 | XP_016866071.1 | 8 | 6204 | |||
XM_017010583.1 | XP_016866072.1 | 9 | 3801 | |||
XM_017010584.1 | XP_016866073.1 | 8 | 8177 | |||
GenBank mRNA | AK023626.1 | - | 1 | 1063 | ||
AK127835.1 | - | 1 | 2114 | |||
AL832366.1 | - | 1 | 1632 | |||
BX537746.1 | - | 8 | 3336 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
