siRNA Details
Gene Symbol: GLRA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024452.2 | NP_001019623.2 | 4 | 869 | ||
NM_001172285.1 | NP_001165756.1 | 4 | 869 | |||
GenBank mRNA | AK309010.1 | - | 4 | 869 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
