siRNA Details
Gene Symbol: MACROD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033087.1 | NP_001028259.1 | 14 | 1074 | ||
NM_080676.5 | NP_542407.2 | 17 | 1512 | |||
XM_017027675.1 | XP_016883164.1 | 11 | 718 | |||
XM_017027676.1 | XP_016883165.1 | 10 | 649 | |||
XM_017027677.1 | XP_016883166.1 | 18 | 1605 | |||
GenBank mRNA | AK125899.1 | - | 10 | 739 | ||
AK131348.1 | - | 17 | 1642 | |||
BC018687.1 | - | 14 | 1077 | 4717477 | ||
BC035876.1 | - | 6 | 515 | |||
EF159161.1 | - | 1317 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
