siRNA Details
Gene Symbol: MACROD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033087.1 | NP_001028259.1 | 13 | 1003 | ||
XM_017027675.1 | XP_016883164.1 | 10 | 647 | |||
XM_017027677.1 | XP_016883166.1 | 17 | 1534 | |||
GenBank mRNA | BC018687.1 | - | 13 | 1006 | 4717477 | |
BC035876.1 | - | 5 | 447 | |||
EF159161.1 | - | 1246 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
