siRNA Details
Gene Symbol: DLEU2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002612.1 | 5, 6 | 594 | |||
GenBank mRNA | AF380424.1 | - | 5, 6 | 773 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
