siRNA Details
Gene Symbol: DLEU2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_002612.1 | 7 | 950 | |||
GenBank mRNA | AF380424.1 | - | 7 | 1129 | ||
AY455757.1 | - | LIT3420 DLEU2 |
10 | 1250 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
