siRNA Details
Gene Symbol: KCTD21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029859.2 | NP_001025030.1 | 2 | 3366 | ||
XM_005273925.3 | XP_005273982.1 | 2 | 3591 | |||
XM_005273928.1 | XP_005273985.1 | 3 | 3491 | |||
XM_006718516.3 | XP_006718579.1 | 2 | 6536 | |||
XM_006718517.2 | XP_006718580.1 | 3 | 3603 | |||
XM_006718518.3 | XP_006718581.1 | 2 | 3351 | |||
XM_011544955.2 | XP_011543257.1 | 2 | 4831 | |||
GenBank mRNA | AK095233.1 | - | 3 | 3115 | ||
BC036058.1 | - | 2 | 3355 | 5277643 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
