siRNA Details
Gene Symbol: TTC24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105669.2 | NP_001099139.2 | 10 | 1670 | ||
XM_011509253.2 | XP_011507555.1 | 9 | 1052 | |||
XM_011509254.2 | XP_011507556.1 | 9 | 1049 | |||
XM_011509255.2 | XP_011507557.1 | 8 | 938 | |||
XM_017000497.1 | XP_016855986.1 | 7 | 674 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
