siRNA Details
Gene Symbol: TTC24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105669.2 | NP_001099139.2 | 3 | 761 | ||
XM_011509253.2 | XP_011507555.1 | 2 | 143 | |||
XM_011509254.2 | XP_011507556.1 | 2 | 143 | |||
XM_011509255.2 | XP_011507557.1 | 2 | 143 | |||
GenBank mRNA | BQ216668.1 | - | 1 | 25 | ||
BX414790.2 | - | 1 | 23 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
