siRNA Details
Gene Symbol: IFITM10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170820.3 | NP_001164291.2 | 2 | 639 | ||
GenBank mRNA | AK289574.1 | - | 1 | 249 | ||
BQ669640.1 | - | 2 | 525 | |||
BQ672240.1 | - | 1 | 219 | |||
BU169250.1 | - | 1 | 407 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
